ENA402267491 Enamine

3-fluoro-4-[1-(propan-2-yl)-1H-1,2,3,4-tetrazol-5-yl]aniline

  • MDL number MFCD16084713
  • Cas Number : 1250227-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118...120
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131892

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131892
Availability details for
Quantity
  • Availability
  • Add to Cart