ENA402267492 Enamine

4-bromo-N-(1H-imidazol-2-ylmethyl)-2,6-dimethylaniline

  • MDL number MFCD14693665
  • Cas Number : 1247827-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131895

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131895
Availability details for
Quantity
  • Availability
  • Add to Cart