ENA402267495 Enamine

1-methyl-2,3-dihydro-1H-indole-6-carboxylic acid

  • MDL number MFCD20660457
  • Cas Number : 1071432-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131901
Availability details for
Quantity
  • Availability
  • Add to Cart