ENA402267499 Enamine

4-(3-chlorophenyl)-4-hydroxycyclohexan-1-one

  • MDL number MFCD21321147
  • Cas Number : 1461708-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131907
Availability details for
Quantity
  • Availability
  • Add to Cart