ENA402267501 Enamine

2-methyl-3-(1-methyl-1H-pyrazol-5-yl)propanal

  • MDL number MFCD20385171
  • Cas Number : 1344347-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131910
Availability details for
Quantity
  • Availability
  • Add to Cart