ENA402267506 Enamine

4-methyl-1-nitro-1H-pyrazole

  • MDL number MFCD03198976
  • Cas Number : 38858-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131916
Availability details for
Quantity
  • Availability
  • Add to Cart