ENA402267507 Enamine

2-[4-(trifluoromethyl)cyclohexyl]acetic acid

  • MDL number MFCD15526995
  • Cas Number : 803736-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131917
Availability details for
Quantity
  • Availability
  • Add to Cart