ENA402267515 Enamine

5-(propan-2-yl)-4H,5H,6H,7H-[1,3]thiazolo[5,4-c]pyridine

  • MDL number MFCD25371598
  • Cas Number : 1461705-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129267
Availability details for
Quantity
  • Availability
  • Add to Cart