ENA402267517 Enamine

1-(4-amino-2-chlorophenyl)ethan-1-one

  • MDL number MFCD11848240
  • Cas Number : 72531-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58...60
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132003

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132003
Availability details for
Quantity
  • Availability
  • Add to Cart