ENA402267519 Enamine

5-(propan-2-yl)-4H,5H,6H,7H-[1,3]thiazolo[5,4-c]pyridin-2-amine

  • MDL number MFCD02854750
  • Cas Number : 17899-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129268
Availability details for
Quantity
  • Availability
  • Add to Cart