ENA402267521 Enamine

5-cyclopropyl-3-methyl-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD11584321
  • Cas Number : 1082329-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132015
Availability details for
Quantity
  • Availability
  • Add to Cart