ENA402267525 Enamine

3-methyl-4-(morpholin-4-yl)aniline

  • MDL number MFCD09755865
  • Cas Number : 112900-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132026

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132026
Availability details for
Quantity
  • Availability
  • Add to Cart