ENA402267526 Enamine

1-[3,5-bis(trifluoromethyl)phenyl]-2,2,2-trifluoroethan-1-amine

  • MDL number MFCD18702245
  • Cas Number : 1270476-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132027
Availability details for
Quantity
  • Availability
  • Add to Cart