ENA402267529 Enamine

2-[2-(propan-2-yl)phenoxy]benzoic acid

  • MDL number MFCD12615716
  • Cas Number : 1036554-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133...135
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132032

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132032
Availability details for
Quantity
  • Availability
  • Add to Cart