ENA402267533 Enamine

3-(difluoromethyl)-4-methyl-4H-1,2,4-triazole

  • MDL number MFCD24197265
  • Cas Number : 1461706-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129284
Availability details for
Quantity
  • Availability
  • Add to Cart