ENA402267534 Enamine

2-{[3-(3-chlorophenyl)-1,2,4-oxadiazol-5-yl]sulfanyl}acetic acid

  • MDL number MFCD17412558
  • Cas Number : 338746-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129327
Availability details for
Quantity
  • Availability
  • Add to Cart