ENA402267538 Enamine

4-(propan-2-yloxy)butanenitrile

  • MDL number MFCD09938370
  • Cas Number : 33563-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132052

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132052
Availability details for
Quantity
  • Availability
  • Add to Cart