ENA402267542 Enamine

3-bromo-N-(4-methoxyphenyl)propanamide

  • MDL number MFCD00563375
  • Cas Number : 7661-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129366

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129366
Availability details for
Quantity
  • Availability
  • Add to Cart