ENA402267545 Enamine

7-chloro-3-methyl-2,3,4,5-tetrahydro-1,5-benzothiazepin-4-one

  • MDL number MFCD25371766
  • Cas Number : 1461708-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222...224
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132061

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132061
Availability details for
Quantity
  • Availability
  • Add to Cart