ENA402267547 Enamine

ethyl 2-[(4-{[(tert-butoxy)carbonyl]amino}phenyl)methyl]-1,3-thiazole-4-carboxylate

  • MDL number MFCD25371767
  • Cas Number : 1461708-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132063

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132063
Availability details for
Quantity
  • Availability
  • Add to Cart