ENA402267549 Enamine

2-(4-aminopiperidin-4-yl)acetic acid dihydrochloride

  • MDL number MFCD25371600
  • Cas Number : 1461709-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 269...271
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129367
Availability details for
Quantity
  • Availability
  • Add to Cart