ENA402267554 Enamine

4-cyanopyridine-3-carboxylic acid

  • MDL number MFCD18256027
  • Cas Number : 827616-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132107

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132107
Availability details for
Quantity
  • Availability
  • Add to Cart