ENA402267558 Enamine

1-(3-methoxypyridin-4-yl)piperazine

  • MDL number MFCD18261788
  • Cas Number : 151140-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132121
Availability details for
Quantity
  • Availability
  • Add to Cart