ENA402267559 Enamine

ethyl 2-(3-bromo-4-methoxyphenyl)-2-hydroxyacetate

  • MDL number MFCD16723848
  • Cas Number : 1247079-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-132124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132124
Availability details for
Quantity
  • Availability
  • Add to Cart