ENA402267570 Enamine

4-(thiophen-3-yl)butanoic acid

  • MDL number MFCD08064092
  • Cas Number : 1505-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132142
Availability details for
Quantity
  • Availability
  • Add to Cart