ENA402267572 Enamine

4-methoxy-1-(2-nitrophenyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD09945370
  • Cas Number : 1485646-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129435
Availability details for
Quantity
  • Availability
  • Add to Cart