ENA402267573 Enamine

6-bromo-2,3,4,5-tetrahydro-1H-1-benzazepin-2-one

  • MDL number MFCD16619138
  • Cas Number : 6729-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132147
Availability details for
Quantity
  • Availability
  • Add to Cart