ENA402267574 Enamine

3-(4-chlorophenyl)propanoyl chloride

  • MDL number MFCD08460401
  • Cas Number : 52085-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132148

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132148
Availability details for
Quantity
  • Availability
  • Add to Cart