ENA402267575 Enamine

3-(dimethylamino)pyridine-4-carbonitrile

  • MDL number MFCD24158794
  • Cas Number : 45865-67-6
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-129469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129469
Availability details for
Quantity
  • Availability
  • Add to Cart