ENA402267580 Enamine

methyl 2-ethyl-3-oxohexanoate

  • MDL number MFCD09954620
  • Cas Number : 68039-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132158
Availability details for
Quantity
  • Availability
  • Add to Cart