ENA402267582 Enamine

6-fluoro-5-methoxy-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD18647833
  • Cas Number : 295779-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132160
Availability details for
Quantity
  • Availability
  • Add to Cart