ENA402267584 Enamine

3-(morpholin-4-yl)pyridine-4-carbonitrile

  • MDL number MFCD24158765
  • Cas Number : 1461707-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129476

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129476
Availability details for
Quantity
  • Availability
  • Add to Cart