ENA402267585 Enamine

2-chloropyrimidine-5-carbaldehyde

  • MDL number MFCD10696891
  • Cas Number : 933702-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104952

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-104952
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart