ENA402267590 Enamine

2-[(3-methoxyphenoxy)methyl]benzoic acid

  • MDL number MFCD09714099
  • Cas Number : 938341-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132172
Availability details for
Quantity
  • Availability
  • Add to Cart