ENA402267595 Enamine

1-(5-methyl-1,3,4-thiadiazol-2-yl)piperidin-4-one

  • MDL number MFCD21132124
  • Cas Number : 1461705-50-9
  • Molecular Weight : 197.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(5-methyl-1,3,4-thiadiazol-2-yl)-4-piperidinone
Inchi Code 1S/C8H11N3OS/c1-6-9-10-8(13-6)11-4-2-7(12)3-5-11/h2-5H2,1H3
Inchi Key BKVBQUYBMLXRLW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-132227

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-132227
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart