ENA402267596 Enamine

1-(4-methoxy-2,5-dimethylphenyl)ethan-1-one

  • MDL number MFCD11553435
  • Cas Number : 104174-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132228
Availability details for
Quantity
  • Availability
  • Add to Cart