ENA402267602 Enamine

1-(bromomethyl)-1-(3-methoxypropoxy)cyclopentane

  • MDL number MFCD16680906
  • Cas Number : 1247514-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132236

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132236
Availability details for
Quantity
  • Availability
  • Add to Cart