ENA402267603 Enamine

methyl 2-bromo-3-[4-(trifluoromethyl)phenyl]propanoate

  • MDL number MFCD12911446
  • Cas Number : 1260761-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132261
Availability details for
Quantity
  • Availability
  • Add to Cart