ENA402267606 Enamine

N-(3-aminopyridin-2-yl)acetamide

  • MDL number MFCD18802513
  • Cas Number : 99314-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132265
Availability details for
Quantity
  • Availability
  • Add to Cart