ENA402267607 Enamine

cyclopropylmethyl 5,6-dichloropyridine-3-carboxylate

  • MDL number MFCD18842245
  • Cas Number : 1340556-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-132268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132268
Availability details for
Quantity
  • Availability
  • Add to Cart