ENA402267608 Enamine

methyl 2-amino-4-nitrobenzoate

  • MDL number MFCD00089419
  • Cas Number : 3558-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156...158
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132272

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132272
Availability details for
Quantity
  • Availability
  • Add to Cart