ENA402267610 Enamine

2-{[(4-methylphenyl)methyl]amino}ethan-1-ol

  • MDL number MFCD04557576
  • Cas Number : 40171-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62...64
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132274

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132274
Availability details for
Quantity
  • Availability
  • Add to Cart