ENA402267616 Enamine

1-(6-methoxypyridin-2-yl)propan-2-one

  • MDL number MFCD18367665
  • Cas Number : 1411241-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132291

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132291
Availability details for
Quantity
  • Availability
  • Add to Cart