ENA402267624 Enamine

2-(1,3-dioxolan-2-yl)-1,3-thiazole

  • MDL number MFCD06209531
  • Cas Number : 24295-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132302
Availability details for
Quantity
  • Availability
  • Add to Cart