ENA402267639 Enamine

6-bromo-4-chloro-8-fluoro-2-(trifluoromethyl)quinoline

  • MDL number MFCD12115098
  • Cas Number : 1156277-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132338
Availability details for
Quantity
  • Availability
  • Add to Cart