ENA402267645 Enamine

3-(2-chloroethyl)pyridine hydrochloride

  • MDL number MFCD01691966
  • Cas Number : 4226-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132347
Availability details for
Quantity
  • Availability
  • Add to Cart