ENA402267646 Enamine

3-(5-chlorothiophen-2-yl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD12137290
  • Cas Number : 1153983-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151...153
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132348

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132348
Availability details for
Quantity
  • Availability
  • Add to Cart