ENA402267647 Enamine

[1-(difluoromethyl)-3,5-dimethyl-1H-pyrazol-4-yl]methanol

  • MDL number MFCD17259773
  • Cas Number : 1341983-71-8
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(difluoromethyl)-3,5-dimethyl-1H-pyrazol-4-yl]methanol
Inchi Code 1S/C7H10F2N2O/c1-4-6(3-12)5(2)11(10-4)7(8)9/h7,12H,3H2,1-2H3
Inchi Key VWXIGHIWQRKXRU-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-132352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132352
Availability details for
Quantity
  • Availability
  • Add to Cart