ENA402267652 Enamine

4-(3,4-dimethylphenyl)butan-1-ol

  • MDL number MFCD11592105
  • Cas Number : 1082435-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132374

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132374
Availability details for
Quantity
  • Availability
  • Add to Cart