ENA402267662 Enamine

1,2-dimethylpiperidin-3-amine

  • MDL number MFCD18975516
  • Cas Number : 1461708-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132388

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132388
Availability details for
Quantity
  • Availability
  • Add to Cart